Learn More
AdipoGen Cytochalasin H
SDP
Supplier:  AdipoGen BVT0447M001
Encompass_Preferred
Chemical. CAS 53760-19-3. Formula C30H39NO5. MW 493.6. Isolated from Phomopsis sp. Potent mycotoxin. Phytotoxin. Actin polymerization inhibitor. Used in actin polymerization and cytoskeletal reorganization studies. Antibacterial, antifungal, nematocidal and antitumor compound. Apoptosis inducer. Anti-angiogenic agent. Shows immunosuppressive activity. Shown to modulate the CNS with potential anti-parkinson activity.
Catalog No. 50-256-1917