Learn More
Cayman Chemical Cytochalasin J, Purity: ≥95%, CAS: 53760-20-6, 1 mg
SDP
Catalog No. 502966370
Encompass_Preferred
A deacetylated analog of cytochalasin H that weakly inhibits actin assembly but significantly alters mitotic spindle microtubule organization and kinetochore structure; suppresses the production of reactive oxygen species by stimulated human neutrophils
Catalog No. 50-296-6370 Supplier Cayman Chemical Supplier No. 184431 MG
A deacetylated analog of cytochalasin H that weakly inhibits actin assembly but significantly alters mitotic spindle microtubule organization and kinetochore structure; suppresses the production of reactive oxygen species by stimulated human neutrophils