Learn More
AdipoGen Cytochalasin J
SDP
Supplier:  AdipoGen BVT0450M001
Encompass_Preferred
Chemical. CAS 53760-20-6. Formula C28H37NO4. MW 451.6. Isolated from Phomopsis sp. Mycotoxin. Phytotoxic and cytotoxic agent. Immunosuppressive activity. Affects chromosome congression and spindle microtubule organization in PtK1 cells. Actin and myosin inhibitor.
Catalog No. 50-256-1913