Learn More
Medchemexpress LLC Cytochrome c (bovine heart) | 9007-43-6 | MFCD00130890 | 95.0% | 12,327 Da | 200MG
SDP
Catalog No. 5000235560
Encompass_Preferred
Cytochrome c (bovine heart) is a heme-containing mitochondrial protein provided as a lyophilized powder for research use. It functions in electron transport and is commonly used as a reagent or standard in biochemical assays of oxidative phosphorylation, electron-transfer reactions, and apoptosis research. The material is characterized by its protein sequence and heme prosthetic group and is supplied at ~95% purity.
  • Heme-containing mitochondrial protein suitable for biochemical studies.
  • Commonly used as a reagent or standard in electron transport and apoptosis assays.
  • Provided as a lyophilized powder for convenient storage and handling.
  • Purity approximately 95.0% as listed by the manufacturer.
  • Available in multiple pack sizes, including a 200 mg pack.
  • Identified by CAS number 9007-43-6 for reference and traceability.
Catalog No. 50-002-35560 Supplier Medchemexpress LLC Supplier No. HY125857C200MG
Cytochrome c (bovine heart) is a heme-containing mitochondrial protein provided as a lyophilized powder for research use. It functions in electron transport and is commonly used as a reagent or standard in biochemical assays of oxidative phosphorylation, electron-transfer reactions, and apoptosis research. The material is characterized by its protein sequence and heme prosthetic group and is supplied at ~95% purity.
  • Heme-containing mitochondrial protein suitable for biochemical studies.
  • Commonly used as a reagent or standard in electron transport and apoptosis assays.
  • Provided as a lyophilized powder for convenient storage and handling.
  • Purity approximately 95.0% as listed by the manufacturer.
  • Available in multiple pack sizes, including a 200 mg pack.
  • Identified by CAS number 9007-43-6 for reference and traceability.