Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Cytochrome C Digestion Standard, 13 nmol, Solid, 1 pk

Catalog No. 50466294 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This digestion standard, created by digesting bovine heart cytochrome C with sequencing grade trypsin, is suitable for evaluating and benchmarking HPLC, UHPLC, and UPLC columns or assessing mass spectrometry and chromatography instrumentation performance.

  • Contains 13 nmol Cytochrome C tryptic digest.
  • Formulation: solid.
  • Separation mode: reversed phase.
  • Detection method: UV.
  • System type: LC.
  • Application: peptide.
  • Lyophilized powder stable for 60 months at room temperature.
  • Reconstituted solution stable 1 week at 8°C; freezing extends stability.
  • Recommended dilution for LC column evaluation: 200 μL of 0.055% trifluoroacetic acid in water.
  • Non-hazardous peptides, exempt from Hazard Communication Standard.
  • For laboratory use only.

Catalog No. 50-466-294 Supplier Waters Corp Supplier No. 186006371
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This digestion standard, created by digesting bovine heart cytochrome C with sequencing grade trypsin, is suitable for evaluating and benchmarking HPLC, UHPLC, and UPLC columns or assessing mass spectrometry and chromatography instrumentation performance.

  • Contains 13 nmol Cytochrome C tryptic digest.
  • Formulation: solid.
  • Separation mode: reversed phase.
  • Detection method: UV.
  • System type: LC.
  • Application: peptide.
  • Lyophilized powder stable for 60 months at room temperature.
  • Reconstituted solution stable 1 week at 8°C; freezing extends stability.
  • Recommended dilution for LC column evaluation: 200 μL of 0.055% trifluoroacetic acid in water.
  • Non-hazardous peptides, exempt from Hazard Communication Standard.
  • For laboratory use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.