Learn More
CPC Scientific H-Lys-Ala-Glu-Arg-Ala-Asp-Leu-Ile-Ala-Tyr-Leu-Lys-Gln-Ala-Thr-Ala-Lys-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific CYTC004B
Encompass
An antigenic sequence used to study the T-cell response.ONE-LETTER SEQUENCE: KAERADLIAYLKQATAKMOLECULAR FORMULA: C84H144N24O25MOLECULAR WEIGHT:1890.21STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [86579-06-8], RESEARCH AREA: ImmunomodulationREFERENCES: J.E.McCormack et al., Proc. Natl. Acad. Sci. USA, 91, 2086 (1994); R.Wang et al., Proc. Natl. Acad. Sci. USA, 95, 3804 (1998); J.D.Bui et al., Cell, 100, 457 (2000); F.Bouzahzah et al., J. Immunol., 170, 741 (2003)
Catalog No. 50-210-9183