Learn More
Sigma Aldrich Fine Chemicals Biosciences Cytochrome c from equine heart, 9007-43-6, MFCD00130890, 50 mg
Catalog No. NC1549917
Encompass
≥95% (SDS-PAGE), Synonym: Cytochrome c from horse heart. The specific sites and extent of oxidation in horse cytochrome c treated with H2O2 and UV were characterized. It was suggested that the state of these sites could be used as a biomarker for the oxidative environment in a cell.
Catalog No. NC1549917 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C250650MG
≥95% (SDS-PAGE), Synonym: Cytochrome c from horse heart. The specific sites and extent of oxidation in horse cytochrome c treated with H2O2 and UV were characterized. It was suggested that the state of these sites could be used as a biomarker for the oxidative environment in a cell.