Learn More
Apexbio Technology LLC Cytochrome c fragment (93-108) 1mg
SDP
Catalog No. 5000094261
Encompass_Preferred
Cytochrome c fragment 93-108 is a peptide derived from cytochrome c a mitochondrial protein involved in apoptosis It is designed to interact with apoptotic protease activating factor-1 (Apaf-1) thereby promoting the assembly of the Apaf-1/caspase-9 apoptosome complex and regulating the caspase activation pathway Cytochrome c fragment 93-108 exerts its biological activity primarily through activation of the caspase cascade specifically the activation of caspase-9 and subsequent caspase-3 Based on these pharmacological properties cytochrome c fragment 93-108 holds research potential in studies of apoptosis-related signaling networks caspase activation pathways and drug screening targeting apoptosis regulators
Catalog No. 50-000-94261 Supplier Apexbio Technology LLC Supplier No. A10621
Cytochrome c fragment 93-108 is a peptide derived from cytochrome c a mitochondrial protein involved in apoptosis It is designed to interact with apoptotic protease activating factor-1 (Apaf-1) thereby promoting the assembly of the Apaf-1/caspase-9 apoptosome complex and regulating the caspase activation pathway Cytochrome c fragment 93-108 exerts its biological activity primarily through activation of the caspase cascade specifically the activation of caspase-9 and subsequent caspase-3 Based on these pharmacological properties cytochrome c fragment 93-108 holds research potential in studies of apoptosis-related signaling networks caspase activation pathways and drug screening targeting apoptosis regulators