Learn More
Enzo Life Sciences Cytochrome c monoclonal antibody (6H2.B4) (100µg)
SDP
Supplier:  Enzo Life Sciences ADI905634100
Encompass_Preferred
Clone: 6H2.B4. Host: Mouse. Isotype: IgG1. Immunogen: Native rat Cytochrome c. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Mouse, Rat. Applications: ELISA, IP. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 0.1% sodium azide. Concentration 1mg/ml. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-6975