Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome C, MP Biomedicals™

Catalog No. MP21014673 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21014673 25 mg
MP21014671 100 mg
MP21014672 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP21014673 Supplier MP Biomedicals, Inc Supplier No. 02101467.3
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Regarded as a universal catalyst of respiration, forming an essential electron-bridge between the respirable substrates and oxygen. Cytochrome C, MP Biomedicals is an electron-carrying mitochondrial protein that plays a vital role in cellular oxidations i

The transition of Cytochrome C between the ferrous and ferric states within the cell makes it an efficient biological electron-transporter.

  • It has been identified as an important mediator in apoptotic pathways.The release of mitochondrial Cytochrome C into the cytoplasm stimulates apoptosis and is commonly used as an indicator of the apoptotic process in the cell.
  • Investigation on the effect of Paris saponin I (PS I) on human gastric carcinoma cell growth (SGC7901 cells) have shown an elevated level cytoplasmic cytochrome c. Results are an inhibition of proliferation in SGC7901 cells by inducing mitochondria-dependent apoptosis through Cytochrome C.
  • The specific sites and extent of oxidation in horse Cytochrome C treated with H2O2 and UV were characterized. It was suggested that the state of these sites could be used as a biomarker for the oxidative environment in a cell.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9007-43-6
Molecular Formula C40H48FeN6O6S4
Molecular Weight (g/mol) 892.94
MDL Number MFCD00130890
InChI Key UMUOLVQYJUIZKD-RHANTFMNSA-L
Synonym cytochrome c, 3-7,12-bis 1-2-amino-3-methylamino-3-oxopropyl sulfanylethyl-18-2-carboxyethyl-3,8,13,17-tetramethylporphyrin-21,23-diid-2-yl propanoic acid; iron 2+
PubChem CID 16057918
SMILES CC(SC(=O)[C@@H](N)CS)C1=C(C)\C2=C\C3=C(C(C)SC(=O)[C@@H](N)CS)C(C)=C4CC5N=C(\C=C6/N([Fe++]N34)\C(=C/C1=N2)C(C)=C6CCC(O)=O)C(CCC(O)=O)=C5C

Specifications

Quantity 25 mg
Solubility Information Dark red to Red-Brown Solution at 10mg plus 1mL of water.
IUPAC Name (1Z,6Z,17Z)-15,19-bis(2-carboxyethyl)-4,9-bis[1-(cysteinylsulfanyl)ethyl]-5,10,14,20-tetramethyl-21,23,24,25-tetraaza-22-ferrahexacyclo[9.9.3.1³,⁶.1¹³,¹⁶.0⁸,²³.0¹⁸,²¹]pentacosa-1,3(25),4,6,8,10,14,16(24),17,19-decaene-22,22-bis(ylium)
Formula Weight ∼12,000
Percent Purity ≥90%
Physical Form Powder
Recommended Storage −20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.