Learn More
TRIBIOSCIENCE INC Cytochrome C Reductase from Porcine Heart, 5 U
SDP
Catalog No. NC3454582
Encompass_Preferred
Tribioscience’s Cytochrome C Reductase from porcine heart is formulated as a crude lyophilized powder containing potassium phosphate, with an approximate pH of 7.0. It is suitable for use in Cytochrome C activity analysis.
Catalog No. NC3454582 Supplier TRIBIOSCIENCE INC Supplier No. TBS21145U
Tribioscience’s Cytochrome C Reductase from porcine heart is formulated as a crude lyophilized powder containing potassium phosphate, with an approximate pH of 7.0. It is suitable for use in Cytochrome C activity analysis.