Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cytochrome P450 17A1 Antibody (YA478) | 57 kDa | 10 UL
SDP

Catalog No. 5000398994
Encompass_Preferred

This Cytochrome P450 17A1 Antibody (YA478) is supplied as a liquid and is suitable for various laboratory applications. It is stable under recommended storage conditions and intended for research use only.

  • Host: Rabbit
  • Reactivity: Human, mouse, rat
  • Clonality number: YA478
  • SwissProt ID: P05093
  • Gene ID: 1586
  • Appearance: Liquid
  • Concentration: 1.0 mg/mL
  • Conjugation: Non-conjugated
  • Application: WB, ICC/IF, IHC-P, FC
  • Dilution ratio: WB: 1:500; ICC/IF: 1:50-1:200; IHC-P: 1:50-1:200; FC: 1:50-1:100
  • Formulation: Supplied in 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol. Preservative: 0.05% Sodium Azide.
  • Storage: Stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles.
  • Shipping: Shipping with blue ice.
  • Molecular weight: 57 kDa

Catalog No. 50-003-98994 Supplier Medchemexpress LLC Supplier No. HYP8044910UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Cytochrome P450 17A1 Antibody (YA478) is supplied as a liquid and is suitable for various laboratory applications. It is stable under recommended storage conditions and intended for research use only.

  • Host: Rabbit
  • Reactivity: Human, mouse, rat
  • Clonality number: YA478
  • SwissProt ID: P05093
  • Gene ID: 1586
  • Appearance: Liquid
  • Concentration: 1.0 mg/mL
  • Conjugation: Non-conjugated
  • Application: WB, ICC/IF, IHC-P, FC
  • Dilution ratio: WB: 1:500; ICC/IF: 1:50-1:200; IHC-P: 1:50-1:200; FC: 1:50-1:100
  • Formulation: Supplied in 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol. Preservative: 0.05% Sodium Azide.
  • Storage: Stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles.
  • Shipping: Shipping with blue ice.
  • Molecular weight: 57 kDa

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.