Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cytochrome P450 1B1 antibody (YA1836) | 10 UL
SDP

Catalog No. 5000440821
Encompass_Preferred

Cytochrome P450 1B1 antibody (YA1836) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting Cytochrome P450 1B1. It is intended for research use only.

  • Rabbit host
  • Recombinant, monoclonal
  • Species reactivity: Human, mouse, rat
  • Immunogen: A synthesized peptide derived from human CYP1B1 aa400-543
  • Isotype: IgG
  • Appearance: Liquid
  • Formulation: Supplied in 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% sodium azide and 0.05% BSA
  • Storage & stability: Stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles
  • Shipping: Shipping with blue ice
  • Applications & dilution ratios: WB (Western Blot): 1:500-1:1000, IHC-P (Immunohistochemistry-Paraffin): 1:50-1:100, FC (Flow Cytometry): 1:50-1:100
  • Sensitivity: Endogenous
  • Conjugation: Non-conjugated
  • Modification: Unmodified

Catalog No. 50-004-40821 Supplier Medchemexpress LLC Supplier No. HYP8209110UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cytochrome P450 1B1 antibody (YA1836) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting Cytochrome P450 1B1. It is intended for research use only.

  • Rabbit host
  • Recombinant, monoclonal
  • Species reactivity: Human, mouse, rat
  • Immunogen: A synthesized peptide derived from human CYP1B1 aa400-543
  • Isotype: IgG
  • Appearance: Liquid
  • Formulation: Supplied in 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% sodium azide and 0.05% BSA
  • Storage & stability: Stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles
  • Shipping: Shipping with blue ice
  • Applications & dilution ratios: WB (Western Blot): 1:500-1:1000, IHC-P (Immunohistochemistry-Paraffin): 1:50-1:100, FC (Flow Cytometry): 1:50-1:100
  • Sensitivity: Endogenous
  • Conjugation: Non-conjugated
  • Modification: Unmodified

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.