Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cytochrome P450 24A1 Antibody (YA2121) | 10 UL
SDP

Catalog No. 5000445206
Encompass_Preferred

Cytochrome P450 24A1 Antibody (YA2121) is a non-conjugated, Rabbit-derived IgG monoclonal antibody, specifically designed to target Cytochrome P450 24A1. It is intended for research use only.

  • Host: Rabbit
  • Clonality: Recombinant, Monoclonal
  • Species reactivity: Human
  • Isotype: IgG
  • Conjugation: Non-conjugated
  • Formulation: Supplied in rabbit IgG in 10mM PBS, pH 7.4, 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide and 50% glycerol.
  • Storage & stability: Stored at -20°C for 1 year, avoiding repeated freeze/thaw cycles.
  • Applications: Western blot (WB) with a dilution ratio of 1:500-1:1000.
  • Function: This cytochrome P450 monooxygenase is crucial for vitamin D catabolism and calcium homeostasis, inactivating calcidiol and calcitriol through C24- and C23-oxidation pathways.
  • Subcellular localization: Mitochondrion

Catalog No. 50-004-45206 Supplier Medchemexpress LLC Supplier No. HYP8237610UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cytochrome P450 24A1 Antibody (YA2121) is a non-conjugated, Rabbit-derived IgG monoclonal antibody, specifically designed to target Cytochrome P450 24A1. It is intended for research use only.

  • Host: Rabbit
  • Clonality: Recombinant, Monoclonal
  • Species reactivity: Human
  • Isotype: IgG
  • Conjugation: Non-conjugated
  • Formulation: Supplied in rabbit IgG in 10mM PBS, pH 7.4, 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide and 50% glycerol.
  • Storage & stability: Stored at -20°C for 1 year, avoiding repeated freeze/thaw cycles.
  • Applications: Western blot (WB) with a dilution ratio of 1:500-1:1000.
  • Function: This cytochrome P450 monooxygenase is crucial for vitamin D catabolism and calcium homeostasis, inactivating calcidiol and calcitriol through C24- and C23-oxidation pathways.
  • Subcellular localization: Mitochondrion

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.