Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cytochrome P450 24A1 Antibody (YA2121) | 50 UL
SDP

Catalog No. 5000445004
Encompass_Preferred

Cytochrome P450 24A1 Antibody (YA2121) is a rabbit-derived, non-conjugated IgG monoclonal antibody specifically targeting Cytochrome P450 24A1. This antibody is intended for research use only. Cytochrome P450 24A1 is a monooxygenase crucial for vitamin D catabolism and calcium homeostasis, inactivating calcidiol and calcitriol via C24- and C23-oxidation pathways.

  • Host: Rabbit
  • Clonality: Monoclonal
  • Conjugation: Non-conjugated
  • Isotype: IgG
  • Appearance: Liquid
  • Purity: Affinity chromatography
  • Application: Western blot (WB)
  • Dilution ratio: 1:500-1:1000
  • Species reactivity: Human
  • Molecular weight: 59 kDa (predicted and observed)
  • Subcellular localization: Mitochondrion
  • Research field: Cardiovascular

Catalog No. 50-004-45004 Supplier Medchemexpress LLC Supplier No. HYP8237650UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cytochrome P450 24A1 Antibody (YA2121) is a rabbit-derived, non-conjugated IgG monoclonal antibody specifically targeting Cytochrome P450 24A1. This antibody is intended for research use only. Cytochrome P450 24A1 is a monooxygenase crucial for vitamin D catabolism and calcium homeostasis, inactivating calcidiol and calcitriol via C24- and C23-oxidation pathways.

  • Host: Rabbit
  • Clonality: Monoclonal
  • Conjugation: Non-conjugated
  • Isotype: IgG
  • Appearance: Liquid
  • Purity: Affinity chromatography
  • Application: Western blot (WB)
  • Dilution ratio: 1:500-1:1000
  • Species reactivity: Human
  • Molecular weight: 59 kDa (predicted and observed)
  • Subcellular localization: Mitochondrion
  • Research field: Cardiovascular

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.