Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cytochrome P450 2C9 Antibody (YA2166) | 100 UL
SDP

Supplier:  Medchemexpress LLC HYP82421100UL

Encompass_Preferred

Cytochrome P450 2C9 Antibody (YA2166) is a Rabbit-derived, non-conjugated IgG monoclonal antibody targeting Cytochrome P450 2C9. It is for research use only and is involved in the metabolism of various endogenous substrates, including fatty acids and steroids, contributing to the pharmacokinetic variability of drugs.

  • Host: Rabbit
  • Clonality: Recombinant, monoclonal
  • Species reactivity: Human
  • Conjugation: Non-conjugated
  • Isotype: IgG
  • Formulation: Supplied in 10mM PBS, pH 7.4, 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide and 50% glycerol
  • Storage: Stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles
  • Shipping: Shipping with blue ice
  • Application & dilution ratio: WB: 1:500-1:1000; ICC/IF: 1:50-1:200; FC: 1:50-1:100

Catalog No. 50-004-45072


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.