Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cytochrome P450 2C9 antibody (YA2166) | 50 UL
SDP

Catalog No. 5000444133
Encompass_Preferred

Cytochrome P450 2C9 Antibody (YA2166) is a Rabbit-derived, non-conjugated IgG monoclonal antibody targeting Cytochrome P450 2C9. It is for research use only.

  • Rabbit-derived IgG monoclonal antibody
  • Targets Cytochrome P450 2C9
  • Reactivity: Human
  • Applications: WB (1:500-1:1000), ICC/IF (1:50-1:200), FC (1:50-1:100)
  • Supplied in 10mM PBS, pH 7.4, 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide and 50% glycerol
  • Storage at -20°C for 1 year, avoid repeated freeze/thaw cycles

Catalog No. 50-004-44133 Supplier Medchemexpress LLC Supplier No. HYP8242150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cytochrome P450 2C9 Antibody (YA2166) is a Rabbit-derived, non-conjugated IgG monoclonal antibody targeting Cytochrome P450 2C9. It is for research use only.

  • Rabbit-derived IgG monoclonal antibody
  • Targets Cytochrome P450 2C9
  • Reactivity: Human
  • Applications: WB (1:500-1:1000), ICC/IF (1:50-1:200), FC (1:50-1:100)
  • Supplied in 10mM PBS, pH 7.4, 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide and 50% glycerol
  • Storage at -20°C for 1 year, avoid repeated freeze/thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.