Learn More
Medchemexpress LLC Cytochrome P450 2E1 Antibody (YA2302) | 10 UL
SDP
Catalog No. 5000443747
Encompass_Preferred
Cytochrome P450 2E1 Antibody (YA2302) is a rabbit-derived, non-conjugated IgG monoclonal antibody that specifically targets Cytochrome P450 2E1. It is designed for research use and has applications in Western Blot (WB) for human, mouse, and rat species. This antibody is formulated in 10mM PBS, pH 7.4, containing 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide, and 50% glycerol.
  • Monoclonal antibody targeting Cytochrome P450 2E1.
  • Suitable for Western Blot (WB) applications.
  • Reacts with human, mouse, and rat samples.
  • Immunogen derived from human CYP2E1 aa52-114.
  • Non-conjugated and unmodified.
Catalog No. 50-004-43747 Supplier Medchemexpress LLC Supplier No. HYP8255710UL
Cytochrome P450 2E1 Antibody (YA2302) is a rabbit-derived, non-conjugated IgG monoclonal antibody that specifically targets Cytochrome P450 2E1. It is designed for research use and has applications in Western Blot (WB) for human, mouse, and rat species. This antibody is formulated in 10mM PBS, pH 7.4, containing 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide, and 50% glycerol.
  • Monoclonal antibody targeting Cytochrome P450 2E1.
  • Suitable for Western Blot (WB) applications.
  • Reacts with human, mouse, and rat samples.
  • Immunogen derived from human CYP2E1 aa52-114.
  • Non-conjugated and unmodified.