Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cytochrome P450 2E1 Antibody (YA2302) | 50 UL
SDP

Catalog No. 5000443744
Encompass_Preferred

Cytochrome P450 2E1 Antibody (YA2302) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting Cytochrome P450 2E1. It is for research use only and demonstrates endogenous sensitivity. This antibody is supplied as a liquid formulation in 10mM PBS, pH 7.4, 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide, and 50% glycerol.

  • Involved in fatty acid metabolism and potentially xenobiotic oxidative metabolism.
  • Catalyzes hydroxylation of carbon-hydrogen bonds, specifically at the omega-1 position of saturated fatty acids.
  • Subcellular localization includes endoplasmic reticulum membrane, microsome membrane, and mitochondrion inner membrane.
  • Application in western blot (WB) with a dilution ratio of 1:500-1:1000.
  • Species reactivity includes human, mouse, and rat.
  • Immunogen is a synthesized peptide derived from human CYP2E1 aa52-114.

Catalog No. 50-004-43744 Supplier Medchemexpress LLC Supplier No. HYP8255750UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cytochrome P450 2E1 Antibody (YA2302) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting Cytochrome P450 2E1. It is for research use only and demonstrates endogenous sensitivity. This antibody is supplied as a liquid formulation in 10mM PBS, pH 7.4, 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide, and 50% glycerol.

  • Involved in fatty acid metabolism and potentially xenobiotic oxidative metabolism.
  • Catalyzes hydroxylation of carbon-hydrogen bonds, specifically at the omega-1 position of saturated fatty acids.
  • Subcellular localization includes endoplasmic reticulum membrane, microsome membrane, and mitochondrion inner membrane.
  • Application in western blot (WB) with a dilution ratio of 1:500-1:1000.
  • Species reactivity includes human, mouse, and rat.
  • Immunogen is a synthesized peptide derived from human CYP2E1 aa52-114.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.