Learn More
Cusabio Technology LLC Cytochrome P450 2E1 CYP2E1
Catalog No. 5000050953
Encompass_Preferred
The Human Cytochrome P450 2E1 (CYP2E1) ELISA Kit is designed to quantitatively measure CYP2E1 levels in serum plasma tissue homogenates and cell lysates using a Sandwich ELISA method This kit detects CYP2E1 with a sensitivity of 10 94 pg/mL and offers a broad detection range from 43 75 pg/mL to 2800 pg/mL ensuring accurate measurement across diverse sample concentrations The assay requires a sample volume of 50-100 L and delivers results within 1-5 hours depending on protocol optimization Absorbance is measured at 450 nm to determine CYP2E1 concentration providing a reliable and standardized approach for research applications Its robust design accommodates various biological matrices making it suitable for studies focused on drug metabolism toxicology or CYP2E1-related disease mechanisms
Catalog No. 50-000-50953 Supplier Cusabio Technology LLC Supplier No. CSBEL006425HU
The Human Cytochrome P450 2E1 (CYP2E1) ELISA Kit is designed to quantitatively measure CYP2E1 levels in serum plasma tissue homogenates and cell lysates using a Sandwich ELISA method This kit detects CYP2E1 with a sensitivity of 10 94 pg/mL and offers a broad detection range from 43 75 pg/mL to 2800 pg/mL ensuring accurate measurement across diverse sample concentrations The assay requires a sample volume of 50-100 L and delivers results within 1-5 hours depending on protocol optimization Absorbance is measured at 450 nm to determine CYP2E1 concentration providing a reliable and standardized approach for research applications Its robust design accommodates various biological matrices making it suitable for studies focused on drug metabolism toxicology or CYP2E1-related disease mechanisms