Learn More
Cusabio Technology LLC cytochrome P450 2E1CYP2E1
Catalog No. 5000049227
Encompass_Preferred
This kit detects CYP2E1 in rat serum plasma and tissue homogenates It offers a detection range of 0 156 ng/mL to 10 ng/mL with a sensitivity of 0 039 ng/mL enabling quantitative measurement using a Sandwich ELISA method The assay can be completed within 1-5 hours requires 50-100 L of sample per test and measures absorbance at 450 nm for reliable results This kit is suitable for research applications requiring precise quantification of CYP2E1 levels in various biological matrices
Catalog No. 50-000-49227 Supplier Cusabio Technology LLC Supplier No. CSBE09782R
This kit detects CYP2E1 in rat serum plasma and tissue homogenates It offers a detection range of 0 156 ng/mL to 10 ng/mL with a sensitivity of 0 039 ng/mL enabling quantitative measurement using a Sandwich ELISA method The assay can be completed within 1-5 hours requires 50-100 L of sample per test and measures absorbance at 450 nm for reliable results This kit is suitable for research applications requiring precise quantification of CYP2E1 levels in various biological matrices