Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cytochrome P450 3A4 Antibody (YA3715) | 10 UL
SDP

Catalog No. 5000444879
Encompass_Preferred

The CYP3A4 protein, a cytochrome P450 monooxygenase, is intricately involved in the metabolism of a diverse array of substrates, including sterols, steroid hormones, retinoids, and fatty acids. It plays a crucial role in various biological processes such as androgen and cholesterol metabolism, retinoid metabolism, and the oxidative metabolism of xenobiotics, acting as a major contributor to drug metabolism. This antibody also contributes to vitamin D catabolism and calcium homeostasis.

  • Mouse-derived and non-conjugated IgG1 monoclonal antibody
  • Targets Cytochrome P450 3A4
  • Reactivity: Human, mouse, rat
  • Formulated in PBS, pH 7.4
  • Store at -20°C for 1 year
  • Avoid repeated freeze / thaw cycles
  • Liquid appearance
  • Suitable for WB, IHC-P, ICC/IF, FC, ELISA applications

Catalog No. 50-004-44879 Supplier Medchemexpress LLC Supplier No. HYP8401810UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The CYP3A4 protein, a cytochrome P450 monooxygenase, is intricately involved in the metabolism of a diverse array of substrates, including sterols, steroid hormones, retinoids, and fatty acids. It plays a crucial role in various biological processes such as androgen and cholesterol metabolism, retinoid metabolism, and the oxidative metabolism of xenobiotics, acting as a major contributor to drug metabolism. This antibody also contributes to vitamin D catabolism and calcium homeostasis.

  • Mouse-derived and non-conjugated IgG1 monoclonal antibody
  • Targets Cytochrome P450 3A4
  • Reactivity: Human, mouse, rat
  • Formulated in PBS, pH 7.4
  • Store at -20°C for 1 year
  • Avoid repeated freeze / thaw cycles
  • Liquid appearance
  • Suitable for WB, IHC-P, ICC/IF, FC, ELISA applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.