Learn More
Apexbio Technology LLC Cytochrome P450 CYP1B1 (190-198) [Homo sapiens] 25mg
SDP
Catalog No. 5000094259
Encompass_Preferred
Cytochrome P450 CYP1B1 190-198 is a tumor-associated antigen peptide derived from CYP1B1 It is designed to induce HLA-A2-restricted T-cell responses against the CYP1B1(190-198) epitope thereby promoting tumor-specific cytotoxic immune responses Cytochrome P450 CYP1B1 190-198 exerts its biological activity primarily through antigen-specific activation of CD8 T cells In HLA-A2 transgenic mouse models this peptide delivered via plasmid DNA and poly(lactide-co-glycolide) microparticles demonstrates the ability to elicit CYP1B1 peptide-specific T-cell immunity Based on these pharmacological properties Cytochrome P450 CYP1B1 190-198 holds research potential in cancer immunotherapy and studies of tumor-specific cytotoxic T-cell responses
Catalog No. 50-000-94259 Supplier Apexbio Technology LLC Supplier No. A106325
Cytochrome P450 CYP1B1 190-198 is a tumor-associated antigen peptide derived from CYP1B1 It is designed to induce HLA-A2-restricted T-cell responses against the CYP1B1(190-198) epitope thereby promoting tumor-specific cytotoxic immune responses Cytochrome P450 CYP1B1 190-198 exerts its biological activity primarily through antigen-specific activation of CD8 T cells In HLA-A2 transgenic mouse models this peptide delivered via plasmid DNA and poly(lactide-co-glycolide) microparticles demonstrates the ability to elicit CYP1B1 peptide-specific T-cell immunity Based on these pharmacological properties Cytochrome P450 CYP1B1 190-198 holds research potential in cancer immunotherapy and studies of tumor-specific cytotoxic T-cell responses