Learn More
Enzo Life Sciences Cytochrome P450 polyclonal antibody (50µg)
SDP
Supplier:  Enzo Life Sciences ADIMFO100D
Encompass_Preferred
Host: Rabbit. Immunogen: Native rat Cytochrome p450. UniProt ID: P05182. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Rabbit, Monkey, Horse, Guinea pig, Dog, Clam. Applications: WB. Recommended Dilutions: Western Blot (1:1,000) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7344