Learn More
Medchemexpress LLC Cytochrome P450 Reductase Antibody (YA2619) | 10 UL
SDP
Catalog No. 5000443939
Encompass_Preferred
The Cytochrome P450 Reductase Antibody (YA2619) is a rabbit-derived, non-conjugated IgG monoclonal antibody. It targets Cytochrome P450 Reductase, an enzyme crucial for electron transfer from NADP to cytochrome P450 in microsomes, and can also supply electrons to heme oxygenase and cytochrome B5. This antibody is suitable for various applications including WB, IHC-P, IHC-F, and ICC/IF.
  • Rabbit-derived host
  • Broad reactivity across human, mouse, and rat
  • Non-conjugated IgG monoclonal antibody
  • High purity through affinity purification
  • Supplied as a liquid formulation
  • Targeted detection of 77 kDa Cytochrome P450 Reductase
Catalog No. 50-004-43939 Supplier Medchemexpress LLC Supplier No. HYP8287410UL
The Cytochrome P450 Reductase Antibody (YA2619) is a rabbit-derived, non-conjugated IgG monoclonal antibody. It targets Cytochrome P450 Reductase, an enzyme crucial for electron transfer from NADP to cytochrome P450 in microsomes, and can also supply electrons to heme oxygenase and cytochrome B5. This antibody is suitable for various applications including WB, IHC-P, IHC-F, and ICC/IF.
  • Rabbit-derived host
  • Broad reactivity across human, mouse, and rat
  • Non-conjugated IgG monoclonal antibody
  • High purity through affinity purification
  • Supplied as a liquid formulation
  • Targeted detection of 77 kDa Cytochrome P450 Reductase