Learn More
Electron Microscopy Sciences CytoCool II 11 OZ
SDP
Catalog No. 501902667
Encompass_Preferred
For the Flash freezing of tissue specimens in the Cryostat as well as being used to harden difficult to cut tissue on a microtome during the sectioning. CytoCool II is an aerosol that does not contain any ozone depleting (CFC'S). Controlling the tissue freezing is simply done by varying the finger pressure on the trigger nozzle. This unique feature helps to prevent over cooling of the specimen.
Catalog No. 50-190-2667 Supplier Electron Microscopy Sciences Supplier No. 7259712
For the Flash freezing of tissue specimens in the Cryostat as well as being used to harden difficult to cut tissue on a microtome during the sectioning. CytoCool II is an aerosol that does not contain any ozone depleting (CFC'S). Controlling the tissue freezing is simply done by varying the finger pressure on the trigger nozzle. This unique feature helps to prevent over cooling of the specimen.