Learn More
StatLab CytoDot Pen
SDP
Catalog No. STL9004
Change view
Click to view available options
Color:
Assorted
Blue
Green
Red
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
STL9004 Red
STL9001 Assorted
STL9002 Green
STL9003 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. STL9004 Supplier StatLab Supplier No. 9004
Designed for ink application on glass slides and coverslips after staining.
  • Fine tip cytology markers are permanent, waterproof, and write on almost any surface including glass, plastic, ceramic and metal
  • 4 pens per pack
Specifications
Tip Style Fine
Color Red
Autoclavable Not Autoclavable
Sterility Non-sterile
Disposable Reusable
Latex Free Yes