Learn More
Beckman Coulter CytoFLEX Reagents Starter Kit for CytoFLEX Flow Cytometers (No IR Laser)
Supplier:  Beckman Coulter B55031
Encompass
Starter kit that provides essential maintenance and quality-control reagents for CytoFLEX flow cytometers without an IR laser. The kit bundles ready-to-use daily QC fluorospheres, sheath fluid, cleaning solutions, and sample tubes to support routine startup, cleaning, and instrument QC.
  • Includes ready-to-use daily QC fluorospheres.
  • Includes sheath fluid for instrument operation.
  • Includes instrument cleaning solution for maintenance.
  • Includes decontamination concentrate for flushing and cleaning.
  • Supplies sample tubes for routine sample handling.
Catalog No. NC2881998