Learn More
Beckman Coulter CytoFLEX SRT, Flow Cytometer
Supplier:  Beckman Coulter C72894
Encompass
The Beckman Coulter CytoFLEX SRT is a benchtop flow cytometer designed for high-throughput research applications. It utilizes a violet (405 nm), blue (488 nm), yellow-green (561 nm), and red (638 nm) laser configuration with 13 fluorescence detectors and forward and side scatter to identify and analyze cells and particles. Key features include up to 70,000 events per second processing speed, a plate loader for high-throughput 96 or 384 well plates, and 21 CFR Part 11 compliant software for secure data management. The compact size, high speed, and plate loading make the CytoFLEX SRT well-suited for high-volume flow cytometry analysis in a research setting.
Catalog No. NC2332497