Learn More
Beckman Coulter CytoFLEX SRT, Flow Cytometer
Catalog No. NC2332501
Encompass
The Beckman Coulter CytoFLEX SRT is a benchtop flow cytometer designed for high-throughput research applications. It has 4 lasers and 21 fluorescence detectors, enabling up to 30-color analysis. The fluidics system processes 70,000 events per second with high sensitivity. It comes with CytExpert software for automated analysis. The CytoFLEX SRT is compact, requiring minimal benchtop space. It offers walkaway automation, high-throughput capabilities, and flexibility for a wide range of applications.
Catalog No. NC2332501 Supplier Beckman Coulter Supplier No. C58834
The Beckman Coulter CytoFLEX SRT is a benchtop flow cytometer designed for high-throughput research applications. It has 4 lasers and 21 fluorescence detectors, enabling up to 30-color analysis. The fluidics system processes 70,000 events per second with high sensitivity. It comes with CytExpert software for automated analysis. The CytoFLEX SRT is compact, requiring minimal benchtop space. It offers walkaway automation, high-throughput capabilities, and flexibility for a wide range of applications.