Learn More
Beckman Coulter Cytoflex Shutdown Fluid [12 EA]
Catalog No. NC2018322
Encompass
Sterile, non-pyrogenic flow cytometry shutdown fluid for CYTOFLEX™ prevents salt crystal formation & biological growth in fluidics system during storage with 0.1% sodium azide preservative for CYTOFLEX™ flow cytometer maintenance & research use only.
Catalog No. NC2018322 Supplier Beckman Coulter Supplier No. C52574
Sterile, non-pyrogenic flow cytometry shutdown fluid for CYTOFLEX™ prevents salt crystal formation & biological growth in fluidics system during storage with 0.1% sodium azide preservative for CYTOFLEX™ flow cytometer maintenance & research use only.