Learn More
Beckman Coulter CytoFLEX SRT, Cell Sorter
Catalog No. NC2332498
Encompass
The Beckman Coulter CytoFLEX SRT is a high-speed cell sorter that provides high purity and recovery in a compact benchtop system. It features a 70um nozzle, 4-way sorting, and up to 4 fluorescent channels plus FSC and SSC. The CytoFLEX SRT enables high-speed sorting of up to 30,000 events per second with >90% purity. It is designed for high-throughput applications including stem cell research, immunology, genomics, and drug discovery. The compact size makes it easy to transport and install in standard biosafety cabinets. The CytoFLEX SRT delivers performance comparable to larger, more expensive sorters in an affordable benchtop system.
Catalog No. NC2332498 Supplier Beckman Coulter Supplier No. C71882
To request a quote for this item, please select Request a Quote to reach our dedicated support team.
The Beckman Coulter CytoFLEX SRT is a high-speed cell sorter that provides high purity and recovery in a compact benchtop system. It features a 70um nozzle, 4-way sorting, and up to 4 fluorescent channels plus FSC and SSC. The CytoFLEX SRT enables high-speed sorting of up to 30,000 events per second with >90% purity. It is designed for high-throughput applications including stem cell research, immunology, genomics, and drug discovery. The compact size makes it easy to transport and install in standard biosafety cabinets. The CytoFLEX SRT delivers performance comparable to larger, more expensive sorters in an affordable benchtop system.