Learn More
Beckman Coulter Beckman Coulter CytoFLEX Startup Pack for Systems with Plate Loader (C14907)
Catalog No. NC2752036
Encompass
The CytoFLEX Startup Pack provides reagents and essential accessories to prepare and perform routine quality control on Beckman Coulter CytoFLEX flow cytometers configured with a sample plate loader and without the IR laser. The kit supports initial system setup and ongoing instrument maintenance.
  • Includes ready-to-use daily QC fluorospheres.
  • Includes sheath fluid for instrument operation.
  • Includes FlowClean cleaning solution for routine maintenance.
  • Includes Contrad 70 concentrate for decontamination.
  • Includes 96-well plates for plate-based sampling.
  • Supports setup and routine quality control for systems with a plate loader.
Catalog No. NC2752036 Supplier Beckman Coulter Supplier No. C14907
The CytoFLEX Startup Pack provides reagents and essential accessories to prepare and perform routine quality control on Beckman Coulter CytoFLEX flow cytometers configured with a sample plate loader and without the IR laser. The kit supports initial system setup and ongoing instrument maintenance.
  • Includes ready-to-use daily QC fluorospheres.
  • Includes sheath fluid for instrument operation.
  • Includes FlowClean cleaning solution for routine maintenance.
  • Includes Contrad 70 concentrate for decontamination.
  • Includes 96-well plates for plate-based sampling.
  • Supports setup and routine quality control for systems with a plate loader.