Learn More
Description
- Available in 1 mm, 2 mm, and 4 mm gap sizes
- Available in Green, Pink, and Orange colors
- Supports electric field strengths up to 25.0 kV/cm
- Sterile and single-use
- Tested for cytotoxicity and endotoxin levels
- QR code provides experiment traceability
- Traceable by raw material, manufacturing lot, and sterilization lot
- Supplied in packs of 50
Specifications
Specifications
| Sterility | Sterile |
| Type | Electroporation Cuvette |
| Disposable | Single-use |
| Capacity (Metric) | 80 to 800 μL |
| Material | Polycarbonate |
| Quantity | 50 Pk. |
| Width (Metric) Gap | 4 mm |
| For Use With (Equipment) | Electroporator |
| For Use With (Application) | Cell and Gene Therapy Research, Development, and Manufacturing |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.