Learn More
Enzo Life Sciences Cytokeratin 1 (human) monoclonal antibody (34betaB4) (Ready-to-Use) (6ml)
SDP
Supplier:  Enzo Life Sciences ENZ30904
Encompass_Preferred
Clone: 34βB4. Isotype: IgG1. Immunogen: Keratin solubilized from human stratum corneum. Source: Purified from ascites. Species reactivity: Human. Specificity: 34βB4 is specific for "high molecular weight" cytokeratin 1, 68 kDa, of "complex" epithelium. The antibody reacts with all squamous epithelium and squamous carcinomas. It does not react with adenocarcinomas of either simple epithelium (hepatoma) or ductal origin (breast, pancreas). Applications: IHC (PS). Purity: Affinity purified. Formulation: Liquid. Ready-to-use solution in PBS containing 0.1% sodium azide and 1% BSA.
Catalog No. 50-201-2482