Learn More
Enzo Life Sciences Cytokeratin 14 polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS171A0100
Encompass_Preferred
Isotype: IgG. Immunogen: Peptide sequence derived from the C-terminus of mouse keratin 14. Species reactivity: Human, Mouse, Rat. Applications: ICC, IHC, WB. Purity: Affinity purified. Formulation: Liquid. In phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
Catalog No. 50-201-2723