Learn More
Enzo Life Sciences Cytokeratin 18 monoclonal antibody (4B11) (100µl)
SDP
Supplier:  Enzo Life Sciences BMLCA11380100
Encompass_Preferred
Clone: 4B11. Isotype: IgG1. Immunogen: Cytokeratins isolated from cultured porcine kidney epithelial cells (LLC-PK). Species reactivity: Human, Mouse, Rat Porcine. Specificity: Recognizes cytokeratin 18 in a variety of species including human, mouse, rat and pig. Applications: ICC, WB. Purity: Purified.
Catalog No. 50-201-0357