Learn More
Biotium Cytokeratin 7/17(C-46), CF740 conjugate, 0.1mg/mL
Supplier:  Biotium BNC740949100
Encompass_Preferred
Recognizes intermediate filament proteins (IFP's) of 55 kDa and 46 kDa, which are identified as cytokeratin 7 and Cytokeratin 17 respectively. On formalin-fixed, paraffin embedded sections, C-46 reacts with keratin 7 only. Cytokeratins comprise a diverse group of intermediate filament proteins (IFPs) that are expressed as pairs in both keratinized and non-keratinized epithelial tissue. Cytokeratins play a critical role in differentiation and tissue specialization and function to maintain the overall structural integrity of epithelial cells. Cytokeratins have been found to be useful markers of tissue differentiation, which is directly applicable to the characterization of malignant tumors. Cytokeratin 7 is a basic cytokeratin, which is found in most glandular and transitional epithelia but not in the stratified squamous epithelia. Cytokeratin 7 is expressed in the epithelial cells of ovary, lung, and breast but not of colon, prostate, or gastrointestinal tract. Cytokeratin 17
Catalog No. 50-006-25479