Learn More
Biogenex Laboratories Cytokeratin 8 &18 : 5D3 : 1mL
SDP
Supplier:  Biogenex Laboratories MU131UC
Encompass_Preferred
Carcinomas may be classified precisely by the analysis of their keratin patterns. Clone 5D3 recognizes an epitope restricted to a few members of the cytokeratin subclasses, specifically cytokeratins 8 and 18. This antibody reacts with all simple epithelia including glandular epithelium and ciliated pseudostratified columnar epithelium localized in thyroid, female breast, gastrointestinal and respiratory tract. 5D3 may be a useful marker for demonstrating columnar cell differentiation when studying biphasic differentiation of basal cells of respiratory or intermediate epithelium. It may also be possible to use 5D3 in a panel of antibodies for the histological subclassification of some carcinomas such as lung carcinoma or adenocarcinoma
Catalog No. 50-197-8879