Learn More
Biogenex Laboratories Cytokeratin 8 : C51 : 6mL
SDP
Supplier:  Biogenex Laboratories AM1425M
Encompass_Preferred
Cytokeratin 8 (52 kD) and 18 (45 kD) comprise a cytokeratin pair asmarkers for simple epithelia. In most situations, cytokeratin 8 exists intissues together with cytokeratin 18, but there are exceptions among somenormal and abnormal epithelial cells. Therefore, it is useful to use bothmonoclonal antibodies (8 and 18) in combination with other monoclonalcytokeratin antibodies when studying cytokeratin expression patterns.Clone C-51 is designed for the specific localization of Cytokeratin 8 anddoes not cross-react with human cytokeratin 7, 17, 18, or 19.
Catalog No. 50-197-8819