Learn More
Enzo Life Sciences Cytokeratin 8 (human) monoclonal antibody (35betaH11) (Ready-to-Use) (6ml)
SDP
Supplier:  Enzo Life Sciences ENZ30902
Encompass_Preferred
Clone: 35βH11. Isotype: IgM. Immunogen: Triton/High salt-insoluble material from the human hepatocellular carcinoma cell line Hep3B. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human. Specificity: 35βH11 is specific for "low molecular weight" cytokeratin 8 (52.5 kDa) characteristically found in "simple" epithelium (e.g., hepatocytes, pancreatic acinar cells, absorptive cells of the GI tract, etc.), ductal structures (e.g., of liver and breast), but not in squamous epithelium. The antibody stains carcinomas such as ductal carcinoma of the breast, liver and pancreas, adenocarcinoma (ovary, lung, endometrium) but does not react with squamous cell carcinoma, sarcoma, melanoma or lymphoma. Applications: IHC (PS), WB. Purity: Affinity purified. Formulation: Liquid. Ready-to-use solution in PBS containing 0.1% sodium azide and 1% BSA.
Catalog No. 50-201-2480