Learn More
Diagnostic Biosystems CYTOKERATIN CAM52 Cytokeratin (CAM5.2)
Catalog No. 502407610
Encompass_Preferred
Anti-Cytokeratin (CAM 5.2) reagent has a primary reactivity with human keratin proteins that correspond to Moll’s peptides #7 and #8, Mr 48 and 52 kilodaltons (kd), respectively. Cytokeratin 7 and 8 are present on secretory epithelia of normal human tissue but not onstratified squamous epithelium. Anti-Cytokeratin (CAM 5.2) stains most epithelial-derived tissue, including liver, renal tubular epithelium, and hepatocellular and renal cell carcinomas. Anti-Cytokeratin (CAM 5.2) might not react with some squamous cell carcinomas.
Catalog No. 50-240-7610 Supplier Diagnostic Biosystems Supplier No. PDM181
Anti-Cytokeratin (CAM 5.2) reagent has a primary reactivity with human keratin proteins that correspond to Moll’s peptides #7 and #8, Mr 48 and 52 kilodaltons (kd), respectively. Cytokeratin 7 and 8 are present on secretory epithelia of normal human tissue but not onstratified squamous epithelium. Anti-Cytokeratin (CAM 5.2) stains most epithelial-derived tissue, including liver, renal tubular epithelium, and hepatocellular and renal cell carcinomas. Anti-Cytokeratin (CAM 5.2) might not react with some squamous cell carcinomas.