Learn More
Biogenex Laboratories Cytokeratin Pan : C11 : 6mL
SDP
Supplier:  Biogenex Laboratories AM3575M
Encompass_Preferred
Human keratins are a family of water-insoluble proteins with molecularweights ranging from 40kD - 68kD. They form a part of the cytoskeletonof epithelial cells. This monoclonal cytokeratin antibody can be used todetect cytokeratins 4, 5, 6, 8, 10, 13, and 18 in simple or stratifiedepithelium in most vertebrates including human. It can be used as amarker for carcinoma as well as some special types of tumors which havean epithelial component or differentiation. Cytokeratin antibodies havebeen widely used as markers to differentiate epithelial tumors from nonepithelial tumors
Catalog No. 50-197-8843