Learn More
Diagnostic Biosystems CYTOKERATIN WIDE SPECTRUM Cytokeratin Wide Spectrum
Catalog No. 502407581
Encompass_Preferred
This antibody reacts with cytokeratins of 58, 56, 52, 60, 51, 48 and 68 kDa MW. This antibody is well suited for the staining of a broad spectrum of human keratins.
Catalog No. 50-240-7581 Supplier Diagnostic Biosystems Supplier No. PDR035
This antibody reacts with cytokeratins of 58, 56, 52, 60, 51, 48 and 68 kDa MW. This antibody is well suited for the staining of a broad spectrum of human keratins.