Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ProcartaPlex™ Human Cytokine/Chemokine/Growth Factor 384 well Panel, 45plex

Catalog No. 502576112
Encompass_Preferred
Change view
Click to view available options
Quantity:
384 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-257-6112 384 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-257-6112 Supplier Invitrogen™ Supplier No. EPXTL45012171901
Add to Cart
Add to Cart
Request a Quote

The ProcartaPlex Human Cytokine/Chemokine/Growth Factor 384 well Panel 45plex (lyophilized beads) enables the study of immune function by analyzing 45 protein targets in a single well using Luminex FLEXMAP 3D and INTELLIFLEX instruments.

The ProcartaPlex Human Cytokine/Chemokine/Growth Factor 384 well Panel 45plex (lyophilized beads) enables the study of immune function by analyzing 45 protein targets in a single well using Luminex FLEXMAP 3D and INTELLIFLEX instruments. The 384-well flat-bottom plate contains ready-to-use lyophilized Luminex beads for fewer pipetting steps, less time-to-result, and seamless integration into workflows.

ProcartaPlex preconfigured panels are extensively tested for analyte combinability, interference, and cross-reactivity to provide the highest level of validation and precision. All ProcartaPlex panels are supplied with the necessary reagents to perform the assay.

Target [bead region]:

Th1/Th2: GM-CSF [44], IFN gamma [43], IL-1 beta [18], IL-2 [19], IL-4 [20], IL-5 [21], IL-6 [25], IL-8 [27], IL-12p70 [34], IL-13 [35], IL-18 [66], TNF alpha [45]

Th9/Th17/Th22/Treg: IL-9 [52], IL-10 [28], IL-17A (CTLA-8) [36], IL-21 [72], IL-22 [76], IL-23 [63], IL-27 [14]

Inflammatory cytokines: IFN alpha [48], IL-1 alpha [62], IL-1RA [38], IL-7 [26], IL-15 [65], IL-31 [37], TNF beta [54]

Chemokines: Eotaxin (CCL11) [33], GRO alpha (CXCL1) [61], IP-10 (CXCL10) [22], MCP-1 (CCL2) [51], MIP-1 alpha (CCL3) [12], MIP-1 beta (CCL4) [47], RANTES (CCL5) [42], SDF-1 alpha [13]

Growth factors: BDNF [57], EGF [56], FGF-2 [75], HGF [46], NGF beta [55], PDGF-BB [77], PlGF-1 [29], SCF [39], VEGF-A [78], VEGF-D [53]

About ProcartaPlex 384-well assays for the Luminex platform
ProcartaPlex 384-well immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using Luminex FLEXMAP 3D and INTELLIFLEX instruments. ProcartaPlex 384-well multiplex assays require as little as 6.3 μL of plasma or serum, or 12.5 μL of cell culture supernatant, and just four hours to obtain analyzed results.

Features include:
• 384-well flat bottom plates with ready-to-use lyophilized Luminex beads for fewer pipetting steps, less time-to-result, and seamless integration into workflows
• Reproducible, reliable results—validated as a panel to the highest industry standard, including protein target combinability and cross-reactivity testing
• More results per sample—measure multiple protein targets simultaneously in a single 6.3–12.5 μL sample
• Well-established Luminex technology highly referenced multiplexing platform for protein detection and quantitation

ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 80 protein targets in a single 6.3–12.5 μL sample—from plasma, serum, cell culture supernatants, and other bodily fluids.

The Luminex beads in the ProcartaPlex 384-well assay are internally dyed with precise proportions of red and infrared fluorophores to create spectrally unique signatures that can be identified by the Luminex xMAP detection systems (e.g., FLEXMAP 3D and INTELLIFLEX). Similar to a sandwich ELISA, the ProcartaPlex 384-well assay uses matched antibody pairs to identify the protein of interest. In a multiplexed assay, each spectrally unique bead is labeled with antibodies specific for a single target protein, and bound proteins are identified with biotinylated antibodies and streptavidin–R-phycoerythrin (RPE). The conjugation of protein-specific antibodies to a distinct bead allows for analysis of multiple targets in a single well.

The most significant difference between a ProcartaPlex assay and ELISA is that the capture antibody in the ProcartaPlex assay is conjugated to a bead and not adsorbed to the microplate well, so the ProcartaPlex assay reagents are free-floating in the solution. For detection, the FLEXMAP 3D instrument, for example, contains two lasers, one to distinguish the spectral signature of each bead and the second to quantify the amount of RPE fluorescence, which is proportional to the amount of protein present in the sample. ProcartaPlex multiplex assays can profile more target proteins using significantly less sample in the same time that it takes to perform a traditional sandwich ELISA.

ProcartaPlex multiplex 384-well panels with lyophilized Luminex beads are available for human and mouse.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Multiplex Panel
Quantity 384 Tests
Species Human
Combinability Non-Combinable
Assay Sensitivity See Certificate of Analysis
Assay Range See Certificate of Analysis
Format Multiplex Kit
For Use With (Equipment) FLEXMAP 3 D™ INTELLIFLEX™
Interassay CV Less than 15%
Intraassay CV Less than 15%
Multiplex Size Plex No. 45
Product Line ProcartaPlex
Sample Type Cell Culture Supernatants, Plasma, Serum
Sample Volume Serum, Plasma: 6.3 μL; CCS: 12.5 μL
Content And Storage

• Human Standard Mix A (lyophilized)
• Human Standard Mix B (lyophilized)
• Human Standard Mix C (lyophilized)
• 384-well plate with lyophilized Capture Bead Mix (1X)
• Biotinylated Detection Antibody Mix (1X)
• Human Control-High A
• Human Control-Low A
• Human Control-High B
• Human Control-Low B
• Human Control-High C
• Human Control-Low C
• Reading Buffer (1X)
• Wash Buffer (10X)
• Streptavidin-PE (1X)
• Universal Assay Buffer (1X)
• 8-tube strip
• Adhesive film

Store at 2°C to 8°C.

Shipping Condition Wet Ice
Research Area Immunology, Cytokines, Chemokines, Growth Factors
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.