Learn More
Electron Microscopy Sciences Single Cytology Funnels, 50/PK
SDP
Catalog No. 502090725
Encompass_Preferred
Fully compatible with the Shandon CytoSpin Centrifuge, the CytoSep Cytology Funnels can be used to deposit a thin layer of cells in a clearly defined area of a microscope slide. The filter card absorbs any access fluid. These Cytology Funnels have the filter cards pre-attached for consistent, reliable results. No alignment necessary. All disposable funnels are packaged with closure caps to seal in specimen for added protection. For sample volumes of up to 0.4ml.
Catalog No. 50-209-0725 Supplier Electron Microscopy Sciences Supplier No. 6275020
Fully compatible with the Shandon CytoSpin Centrifuge, the CytoSep Cytology Funnels can be used to deposit a thin layer of cells in a clearly defined area of a microscope slide. The filter card absorbs any access fluid. These Cytology Funnels have the filter cards pre-attached for consistent, reliable results. No alignment necessary. All disposable funnels are packaged with closure caps to seal in specimen for added protection. For sample volumes of up to 0.4ml.