Learn More
Electron Microscopy Sciences CytoSep™ Cytology Single Funnel Tubing with Cap
SDP
Catalog No. 501900464
Encompass_Preferred
This plastic CytoSep MEGA Funnel provides an easy, efficient and cost effective method of producing high quality thin-layer slide preparations and can be used for preparation of large volume samples. It eliminates the needs for stainless steel clips.
The larger funnel is quick and easy to remove. The large rectangular cell deposition area of 22 x 14.75 mm (325 mm squared) provides for up to 12 times the sample volume (6 ml) of the single Cyto Cytology Funnel. Strategically placed baffles inhibit cell setting which results in uniform cell deposition and excellent quality slide preparations. This MEGA funnels and cap minimize user exposure to pathogens while reducing the risk of specimen cross contamination. Can prepared both air dried and fixed preparations.
Catalog No. 50-190-0464 Supplier Electron Microscopy Sciences Supplier No. 6275410
This plastic CytoSep MEGA Funnel provides an easy, efficient and cost effective method of producing high quality thin-layer slide preparations and can be used for preparation of large volume samples. It eliminates the needs for stainless steel clips.
The larger funnel is quick and easy to remove. The large rectangular cell deposition area of 22 x 14.75 mm (325 mm squared) provides for up to 12 times the sample volume (6 ml) of the single Cyto Cytology Funnel. Strategically placed baffles inhibit cell setting which results in uniform cell deposition and excellent quality slide preparations. This MEGA funnels and cap minimize user exposure to pathogens while reducing the risk of specimen cross contamination. Can prepared both air dried and fixed preparations.