Learn More
Volu Sol Gill's Hematoxylin III (Triple Strength), 128 oz / 3.78 L, Volu-Sol™
SDP
Catalog No. 50420858 Shop All Volu Sol Products
Encompass_Preferred
Nuclear stain, progressive.  Used with hematoxylin and eosin staining
Catalog No. 50-420-858 Supplier Volu Sol Supplier No. VCH128
Nuclear stain, progressive.  Used with hematoxylin and eosin staining