Learn More
Volu Sol Gill's Hematoxylin III (Triple Strength), 32 oz / 1 L, Volu-Sol™
SDP
Catalog No. 50420857 Shop All Volu Sol Products
Encompass_Preferred
Nuclear stain, progressive.  Used with hematoxylin and eosin staining
Catalog No. 50-420-857 Supplier Volu Sol Supplier No. VCH032
Nuclear stain, progressive.  Used with hematoxylin and eosin staining