Learn More
Bio Rad Laboratories Cytometer Cleaner | 1L
Supplier:  Bio Rad Laboratories 12004272
Encompass
Use this cytometer system-cleaning reagent in the ZE5 Cell Analyzer to minimize sample buildup over time and preserve the integrity of the system.
Catalog No. NC1649509